siRNA Details
| Gene Symbol: KDF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152365.2 | NP_689578.2 | 2 | 733 | ||
| XM_005245735.2 | XP_005245792.1 | 2 | 774 | |||
| XM_011540622.1 | XP_011538924.1 | 2 | 748 | |||
| GenBank mRNA | AK091952.1 | - | 2 | 729 | ||
| BC033143.1 | - | 1 | 564 | 3960220 | ||
| CR749558.1 | - | 3 | 590 | |||

KDF1
Genomic Map