siRNA Details
| Gene Symbol: OR2M3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004689.1 | NP_001004689.1 | 1 | 12 | ||
| GenBank mRNA | BC136970.1 | - | 1 | 50 | 9020962 | |
| BC136971.1 | - | 1 | 50 | 9020963 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR2M3| Gene Symbol: OR2M3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004689.1 | NP_001004689.1 | 1 | 12 | ||
| GenBank mRNA | BC136970.1 | - | 1 | 50 | 9020962 | |
| BC136971.1 | - | 1 | 50 | 9020963 | ||