siRNA Details
| Gene Symbol: ASB17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080868.2 | NP_543144.1 | 1 | 508 | ||
| NR_026546.1 | 1 | 508 | ||||
| XM_006710345.3 | XP_006710408.1 | 2 | 340 | |||
| GenBank mRNA | AF403035.1 | - | 1 | 368 | ||
| AF429968.1 | - | 1 | 485 | |||
| AK098606.1 | - | 1 | 508 | |||
| BC022517.1 | - | 1 | 441 | |||
| BC036573.2 | - | 1 | 486 | 5297837 | ||

ASB17
Genomic Map