siRNA Details
| Gene Symbol: TPRG1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182752.3 | NP_877429.2 | 3 | 497 | ||
| GenBank mRNA | AK025464.1 | - | 2 | 217 | ||
| AK289919.1 | - | 3 | 447 | |||
| AK298343.1 | - | 3 | 336 | |||
| BI547609.1 | - | 3 | 492 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TPRG1L| Gene Symbol: TPRG1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182752.3 | NP_877429.2 | 3 | 497 | ||
| GenBank mRNA | AK025464.1 | - | 2 | 217 | ||
| AK289919.1 | - | 3 | 447 | |||
| AK298343.1 | - | 3 | 336 | |||
| BI547609.1 | - | 3 | 492 | |||