siRNA Details
| Gene Symbol: MYOM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152372.3 | NP_689585.3 | 31 | 3806 | ||
| GenBank mRNA | AK093280.1 | - | 4 | 692 | ||
| AK128233.1 | - | 23 | 2866 | |||
| BC014671.1 | - | 5 | 326 | |||
| BX537911.1 | - | 30 | 3530 | |||
| BX647453.1 | - | 31 | 3767 | |||
| BX647455.1 | - | 31 | 3806 | |||
| BX647644.1 | - | 31 | 3767 | |||

MYOM3
Genomic Map