siRNA Details
| Gene Symbol: OR10J5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004469.1 | NP_001004469.1 | 1 | 791 | ||
| GenBank mRNA | BC137025.1 | - | 1 | 836 | 9021020 | |
| BC137026.1 | - | 1 | 836 | 9021021 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR10J5| Gene Symbol: OR10J5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004469.1 | NP_001004469.1 | 1 | 791 | ||
| GenBank mRNA | BC137025.1 | - | 1 | 836 | 9021020 | |
| BC137026.1 | - | 1 | 836 | 9021021 | ||