siRNA Details
| Gene Symbol: LRRC39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256385.1 | NP_001243314.1 | 7 | 823 | ||
| NM_001256386.1 | NP_001243315.1 | 7 | 823 | |||
| NM_001256387.1 | NP_001243316.1 | 6 | 766 | |||
| NM_144620.3 | NP_653221.1 | 7 | 806 | |||
| GenBank mRNA | AK096892.1 | - | 7 | 749 | ||
| AL833417.1 | - | 7 | 771 | |||
| AY358131.1 | - | 6 | 739 | |||
| BC009613.1 | - | 7 | 748 | 4245944 | ||

LRRC39
Genomic Map