siRNA Details
| Gene Symbol: OR10Z1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004478.1 | NP_001004478.1 | 1 | 859 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR10Z1| Gene Symbol: OR10Z1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004478.1 | NP_001004478.1 | 1 | 859 | ||