siRNA Details
| Gene Symbol: OR6K6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005184.1 | NP_001005184.1 | 1 | 549 | ||
| GenBank mRNA | BC140743.1 | - | 1 | 645 | 9021611 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR6K6| Gene Symbol: OR6K6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005184.1 | NP_001005184.1 | 1 | 549 | ||
| GenBank mRNA | BC140743.1 | - | 1 | 645 | 9021611 | |