siRNA Details
| Gene Symbol: TSPY26P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002781.1 | 1 | 747 | |||
| GenBank mRNA | BC101556.1 | - | 1 | 747 | ||
| BC101558.1 | - | 1 | 747 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TSPY26P| Gene Symbol: TSPY26P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002781.1 | 1 | 747 | |||
| GenBank mRNA | BC101556.1 | - | 1 | 747 | ||
| BC101558.1 | - | 1 | 747 | |||