siRNA Details
| Gene Symbol: DIS3L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001257281.1 | NP_001244210.1 | 9 | 1274 | ||
| NM_152383.4 | NP_689596.4 | 9 | 1274 | |||
| NR_046476.1 | 9 | 1274 | ||||
| NR_046477.1 | 8 | 1250 | ||||
| GenBank mRNA | AK094293.1 | - | 9 | 1275 | ||
| AK295363.1 | - | 9 | 1144 | |||
| AL834174.1 | - | 8 | 872 | |||
| BC026166.1 | - | 9 | 1178 | 4822064 | ||
| BC036113.1 | - | 9 | 1173 | 5297794 | ||
| BX648325.1 | - | 9 | 1141 | |||

DIS3L2
Genomic Map