siRNA Details
Gene Symbol: NBEAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114132.1 | NP_001107604.1 | 3, 4 | 486 | ||
XM_005246787.3 | XP_005246844.1 | 3, 4 | 512 | |||
XM_006712698.3 | XP_006712761.1 | 3, 4 | 512 | |||
XM_006712699.3 | XP_006712762.1 | 3, 4 | 512 | |||
XM_011511658.2 | XP_011509960.1 | 3, 4 | 467 | |||
XM_011511659.2 | XP_011509961.1 | 3, 4 | 524 | |||
XM_011511663.2 | XP_011509965.1 | 3, 4 | 502 | |||
GenBank mRNA | AB053318.1 | - | 3, 4 | 400 | ||
AW028696.1 | - | 2 | 240 | |||
BC132950.1 | - | 3, 4 | 440 | |||
BC137214.1 | - | 3, 4 | 440 | |||
BC144532.1 | - | 3, 4 | 440 |
