siRNA Details
| Gene Symbol: PLEKHH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172069.3 | NP_742066.2 | 13 | 2252 | ||
| XM_017003351.1 | XP_016858840.1 | 12 | 2239 | |||
| XM_017003352.1 | XP_016858841.1 | 13 | 2252 | |||
| XM_017003353.1 | XP_016858842.1 | 14 | 2363 | |||
| GenBank mRNA | AB095948.1 | - | 11 | 2038 | ||
| AK126308.1 | - | 13 | 2248 | |||
| AL832207.1 | - | 13 | 2245 | |||
| AL833400.1 | - | 6 | 555 | |||
| AL833546.1 | - | 14 | 2447 | |||
| BC063310.1 | - | 13 | 2279 | 30342490 | ||

PLEKHH2
Genomic Map