siRNA Details
| Gene Symbol: PLEKHH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172069.3 | NP_742066.2 | 15 | 2412 | ||
| XM_017003351.1 | XP_016858840.1 | 14 | 2399 | |||
| XM_017003352.1 | XP_016858841.1 | 15 | 2412 | |||
| XM_017003353.1 | XP_016858842.1 | 16 | 2523 | |||
| GenBank mRNA | AB095948.1 | - | 13 | 2198 | ||
| AK126308.1 | - | 15 | 2408 | |||
| AL832207.1 | - | 15 | 2405 | |||
| AL833400.1 | - | 8 | 715 | |||
| AL833546.1 | - | 15 | 4258 | |||

PLEKHH2
Genomic Map