siRNA Details
| Gene Symbol: GALM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138801.2 | NP_620156.1 | 2 | 466 | ||
| XM_011532540.1 | XP_011530842.1 | 2 | 466 | |||
| GenBank mRNA | AK291489.1 | - | 2 | 274 | ||
| BC014916.1 | - | 2 | 248 | 3920640 | ||
| BC019263.1 | - | 2 | 279 | 3502667 | ||
| DA984992.1 | - | 2 | 466 | |||
| EU794611.1 | - | 2 | 248 | |||
| U11036.1 | - | 2 | 265 | |||

GALM
Genomic Map