siRNA Details
| Gene Symbol: TMEM198 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005209.2 | NP_001005209.1 | 5 | 1429 | ||
| NM_001303098.1 | NP_001290027.1 | 6 | 1603 | |||
| XM_017003367.1 | XP_016858856.1 | 5 | 1385 | |||
| GenBank mRNA | AK123175.1 | - | 3 | 843 | ||
| AK302369.1 | - | 4 | 958 | |||
| BC068567.1 | - | 6 | 1603 | |||
| BC080573.1 | - | 5 | 1412 | 6569597 | ||

TMEM198
Genomic Map