siRNA Details
| Gene Symbol: TMEM178A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167959.1 | NP_001161431.1 | 4 | 424 | ||
| NM_152390.2 | NP_689603.2 | 4 | 761 | |||
| XM_005264144.1 | XP_005264201.1 | 3 | 601 | |||
| XM_005264145.1 | XP_005264202.1 | 2 | 487 | |||
| XM_017003370.1 | XP_016858859.1 | 4 | 543 | |||
| XM_017003371.1 | XP_016858860.1 | 4 | 460 | |||
| GenBank mRNA | AK075440.1 | - | 4 | 737 | ||
| AY358773.1 | - | 4 | 414 | |||
| BC029530.1 | - | 4 | 706 | 5286077 | ||
| BI757636.1 | - | 4 | 423 | |||

TMEM178A
Genomic Map