siRNA Details
| Gene Symbol: MTERF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182501.3 | NP_872307.2 | 2, 3 | 573 | ||
| NR_028049.1 | 2, 3 | 573 | ||||
| XM_006712295.3 | XP_006712358.1 | 2, 3 | 655 | |||
| XM_011510634.2 | XP_011508936.1 | 2, 3 | 830 | |||
| XM_017003387.1 | XP_016858876.1 | 2, 3 | 830 | |||
| XM_017003388.1 | XP_016858877.1 | 2, 3 | 830 | |||
| GenBank mRNA | AK131298.1 | - | 2, 3 | 1347 | ||
| AK291665.1 | - | 2, 3 | 528 | |||
| AK296515.1 | - | 2, 3 | 551 | |||
| BC053533.1 | - | 2, 3 | 526 | 4417008 | ||
| BX648184.1 | - | 2, 3 | 539 | |||

MTERF4
Genomic Map