siRNA Details
| Gene Symbol: ZPLD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175056.1 | NP_778226.1 | 7 | 879 | ||
| XM_017005703.1 | XP_016861192.1 | 8 | 1061 | |||
| XM_017005704.1 | XP_016861193.1 | 7 | 917 | |||
| GenBank mRNA | AY090780.1 | - | 7 | 879 | ||
| BC021279.2 | - | 11 | ||||
| BC143817.1 | - | 8 | 975 | |||

ZPLD1
Genomic Map