siRNA Details
| Gene Symbol: GRK7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139209.2 | NP_631948.1 | 2 | 837 | ||
| XM_017005717.1 | XP_016861206.1 | 4 | 3579 | |||
| GenBank mRNA | AF282269.1 | - | 2 | 837 | ||
| AF439409.1 | - | 2 | 890 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GRK7| Gene Symbol: GRK7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139209.2 | NP_631948.1 | 2 | 837 | ||
| XM_017005717.1 | XP_016861206.1 | 4 | 3579 | |||
| GenBank mRNA | AF282269.1 | - | 2 | 837 | ||
| AF439409.1 | - | 2 | 890 | |||