siRNA Details
Gene Symbol: H1FOO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153833.2 | NP_722575.1 | 1 | 67 | ||
GenBank mRNA | AY158091.1 | - | 1 | 67 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: H1FOO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153833.2 | NP_722575.1 | 1 | 67 | ||
GenBank mRNA | AY158091.1 | - | 1 | 67 |