siRNA Details
| Gene Symbol: JMY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152405.4 | NP_689618.4 | 9 | 2625 | ||
| XM_011543155.2 | XP_011541457.1 | 8 | 2529 | |||
| GenBank mRNA | AK095189.1 | - | 8 | 1026 | ||
| BC130624.1 | - | 8 | 1051 | 40146655 | ||

JMY
Genomic Map