siRNA Details
| Gene Symbol: ZNF474 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207317.2 | NP_997200.1 | 2 | 705 | ||
| GenBank mRNA | AK057483.1 | - | 2 | 699 | ||
| AK290985.1 | - | 2 | 635 | |||
| AY461732.1 | - | 2 | 698 | |||
| BC133022.1 | - | 1 | 380 | 40147044 | ||
| BC133026.1 | - | 1 | 380 | 40147048 | ||
| Gene Symbol: LOC100505841 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017008912.1 | XP_016864401.1 | 2 | 672 | ||

ZNF474
Genomic Map