siRNA Details
| Gene Symbol: OR2Y1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001657.1 | NP_001001657.1 | 1 | 122 | ||
| GenBank mRNA | BC140756.1 | - | 1 | 163 | 9021624 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR2Y1| Gene Symbol: OR2Y1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001657.1 | NP_001001657.1 | 1 | 122 | ||
| GenBank mRNA | BC140756.1 | - | 1 | 163 | 9021624 | |