siRNA Details
| Gene Symbol: AFAP1L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146337.2 | NP_001139809.1 | 11 | 1343 | ||
| NM_001323062.1 | NP_001309991.1 | 10 | 1244 | |||
| NM_152406.3 | NP_689619.1 | 11 | 1343 | |||
| XM_011537558.1 | XP_011535860.1 | 7 | 893 | |||
| XM_017009036.1 | XP_016864525.1 | 11 | 1320 | |||
| GenBank mRNA | AK074185.1 | - | 11 | 1286 | ||
| AK094067.1 | - | 2 | 373 | |||
| AK095980.1 | - | 11 | 1297 | |||
| BC125093.1 | - | 11 | 1262 | 40118258 | ||
| BC125094.1 | - | 11 | 1262 | 40118263 | ||

AFAP1L1
Genomic Map