siRNA Details
| Gene Symbol: AFAP1L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146337.2 | NP_001139809.1 | 6 | 594 | ||
| NM_001323063.1 | NP_001309992.1 | 6 | 594 | |||
| NM_152406.3 | NP_689619.1 | 6 | 594 | |||
| XM_011537558.1 | XP_011535860.1 | 2 | 144 | |||
| XM_017009036.1 | XP_016864525.1 | 6 | 571 | |||
| GenBank mRNA | AK074185.1 | - | 6 | 537 | ||
| AK095980.1 | - | 6 | 548 | |||
| BC040723.2 | - | 6 | 561 | |||
| BC125093.1 | - | 6 | 513 | 40118258 | ||
| BC125094.1 | - | 6 | 513 | 40118263 | ||

AFAP1L1
Genomic Map