siRNA Details
Gene Symbol: TMEM171 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161342.2 | NP_001154814.1 | 4 | 1019 | ||
NM_173490.7 | NP_775761.4 | 4 | 1022 | |||
XM_011543156.1 | XP_011541458.1 | 4 | 945 | |||
GenBank mRNA | BC018083.1 | - | 4 | 1008 | 4797244 | |
BC035310.1 | - | 3 | 860 | 4746772 |
