siRNA Details
| Gene Symbol: POC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099271.1 | NP_001092741.1 | 7 | 946 | ||
| NM_152408.2 | NP_689621.2 | 6 | 745 | |||
| XM_005248436.1 | XP_005248493.1 | 7 | 946 | |||
| XM_011543158.2 | XP_011541460.1 | 7 | 946 | |||
| XM_011543159.1 | XP_011541461.1 | 8 | 952 | |||
| XM_011543160.1 | XP_011541462.1 | 6 | 806 | |||
| XM_017009038.1 | XP_016864527.1 | 6 | 866 | |||
| XM_017009039.1 | XP_016864528.1 | 6 | 785 | |||
| GenBank mRNA | AK093098.1 | - | 6 | 715 | ||
| AK296071.1 | - | 7 | 863 | |||
| BC101325.1 | - | 6 | 711 | 40024984 | ||
| BC101326.2 | - | 6 | 715 | 40024987 | ||
| BC101327.2 | - | 6 | 715 | 40024989 | ||
| BC101328.2 | - | 6 | 712 | |||

POC5
Genomic Map