siRNA Details
| Gene Symbol: WDR36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139281.2 | NP_644810.1 | 8 | 1111 | ||
| GenBank mRNA | AF385437.1 | - | 8 | 1111 | ||
| AK074202.1 | - | 2 | 161 | |||
| AK095312.1 | - | 8 | 969 | |||
| AL832494.1 | - | 7 | 917 | |||
| BC133025.1 | - | 8 | 1071 | 40147047 | ||
| BC136517.1 | - | 8 | 1071 | 9020507 | ||
| CR749211.1 | - | 3 | 311 | |||

WDR36
Genomic Map