siRNA Details
| Gene Symbol: SHROOM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172700.1 | NP_001166171.1 | 7 | 2475 | ||
| NM_133456.2 | NP_597713.2 | 4 | 1840 | |||
| XM_005271885.4 | XP_005271942.1 | 5 | 2841 | |||
| XM_005271886.3 | XP_005271943.1 | 5 | 1753 | |||
| XM_011543167.2 | XP_011541469.1 | 6 | 2235 | |||
| GenBank mRNA | AB075840.1 | - | 7 | 2475 | ||
| AF314142.1 | - | 4 | 1840 | |||
| BC104914.1 | - | 4 | 1702 | 8143917 | ||
| BC104916.1 | - | 4 | 1702 | 8143919 | ||
| BC143494.1 | - | 4 | 1702 | 9052002 | ||

SHROOM1
Genomic Map