siRNA Details
| Gene Symbol: WBSCR28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182504.3 | NP_872310.2 | 2, 3 | 548 | ||
| XM_011515785.2 | XP_011514087.1 | 2, 3 | 482 | |||
| XM_017011741.1 | XP_016867230.1 | 2, 3 | 670 | |||
| GenBank mRNA | AY372053.1 | - | 2, 3 | 553 | ||
| AY372054.1 | - | 3, 4 | 599 | |||
| BC030643.2 | - | 2, 3 | 548 | 4824662 | ||

WBSCR28
Genomic Map