siRNA Details
Gene Symbol: SSMEM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145268.3 | NP_660311.1 | 1, 2 | 229 | ||
GenBank mRNA | AK097635.1 | - | 1, 2 | 215 | ||
BC017587.2 | - | 1, 2 | 233 | 4838989 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: SSMEM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145268.3 | NP_660311.1 | 1, 2 | 229 | ||
GenBank mRNA | AK097635.1 | - | 1, 2 | 215 | ||
BC017587.2 | - | 1, 2 | 233 | 4838989 |