siRNA Details
| Gene Symbol: ASB10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142459.1 | NP_001135931.2 | 1 | 511 | ||
| NM_001142460.1 | NP_001135932.2 | 1 | 511 | |||
| GenBank mRNA | AF417920.1 | - | 1 | 513 | ||
| AK055536.1 | - | 1 | 221 | |||
| AK315114.1 | - | 1 | 341 | |||
| BC126351.1 | - | 1 | 275 | 8992067 | ||

ASB10
Genomic Map