siRNA Details
| Gene Symbol: ASB10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142459.1 | NP_001135931.2 | 2 | 793 | ||
| NM_001142460.1 | NP_001135932.2 | 2 | 793 | |||
| NM_080871.3 | NP_543147.2 | 2 | 597 | |||
| GenBank mRNA | AF417920.1 | - | 2 | 795 | ||
| AK055536.1 | - | 2 | 503 | |||
| AK125565.1 | - | 2 | 597 | |||
| AK315114.1 | - | 2 | 623 | |||
| BC126351.1 | - | 2 | 557 | 8992067 | ||

ASB10
Genomic Map