siRNA Details
| Gene Symbol: ZNF572 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152412.2 | NP_689625.2 | 3 | 424 | ||
| GenBank mRNA | AK095321.1 | - | 3 | 411 | ||
| BC130515.1 | - | 3 | 403 | 40146546 | ||
| BC130517.1 | - | 3 | 403 | 40146548 | ||
| BX537876.1 | - | 3 | 424 | |||
| DB093007.1 | - | 3 | 424 | |||

ZNF572
Genomic Map