siRNA Details
| Gene Symbol: TMEM68 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286657.1 | NP_001273586.1 | 4 | 774 | ||
| NM_001286661.1 | NP_001273590.1 | 3 | 474 | |||
| NM_152417.2 | NP_689630.1 | 5 | 913 | |||
| XM_005251150.1 | XP_005251207.1 | 5 | 902 | |||
| XM_011517454.1 | XP_011515756.1 | 5 | 926 | |||
| XM_011517455.2 | XP_011515757.1 | 5 | 902 | |||
| XM_017013040.1 | XP_016868529.1 | 4 | 486 | |||
| GenBank mRNA | AK056932.1 | - | 5 | 832 | ||
| AL832935.1 | - | 3 | 383 | |||
| CN272366.1 | - | 3 | 401 | |||

TMEM68
Genomic Map