siRNA Details
| Gene Symbol: SGCZ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001322879.1 | NP_001309808.1 | 3 | 913 | ||
| NM_001322880.1 | NP_001309809.1 | 4 | 1015 | |||
| NM_001322881.1 | NP_001309810.1 | 4 | 921 | |||
| NM_139167.3 | NP_631906.2 | 4 | 1015 | |||
| GenBank mRNA | AY028700.1 | - | 3 | 316 | ||
| AY502063.1 | - | 4 | 355 | |||
| BC069459.1 | - | 3 | 316 | |||
| BC125037.1 | - | 2 | 214 | 40114834 | ||

SGCZ
Genomic Map