siRNA Details
Gene Symbol: SGCZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322879.1 | NP_001309808.1 | 2 | 801 | ||
NM_001322880.1 | NP_001309809.1 | 2 | 801 | |||
NM_139167.3 | NP_631906.2 | 2 | 801 | |||
GenBank mRNA | AY028700.1 | - | 1 | 102 | ||
AY502063.1 | - | 2 | 141 | |||
BC069459.1 | - | 1 | 102 | |||
BC125037.1 | - | 1 | 102 | 40114834 |
