siRNA Details
| Gene Symbol: GPAT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178819.3 | NP_848934.1 | 4 | 1202 | ||
| XM_011544392.2 | XP_011542694.1 | 5 | 1325 | |||
| XM_011544393.1 | XP_011542695.1 | 4 | 1214 | |||
| XM_011544394.1 | XP_011542696.1 | 4 | 1251 | |||
| XM_017013042.1 | XP_016868531.1 | 5 | 1352 | |||
| XM_017013043.1 | XP_016868532.1 | 4 | 1200 | |||
| GenBank mRNA | AF317516.1 | - | 3 | 428 | ||
| AF406612.1 | - | 4 | 1190 | |||
| AY358670.1 | - | 3 | 339 | |||
| AY513610.1 | - | 3 | 422 | |||
| BC051377.1 | - | 3 | 382 | 6649895 | ||
| BC061884.1 | - | 3 | 382 | 6577569 | ||

GPAT4
Genomic Map