siRNA Details
| Gene Symbol: CARNMT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001320497.1 | NP_001307426.1 | 5 | 676 | ||
| NM_152420.2 | NP_689633.1 | 5 | 996 | |||
| NR_135282.1 | 4 | 855 | ||||
| XM_005251701.3 | XP_005251758.2 | 5 | 671 | |||
| XM_017014269.1 | XP_016869758.1 | 5 | 983 | |||
| XM_017014270.1 | XP_016869759.1 | 6 | 929 | |||
| XM_017014271.1 | XP_016869760.1 | 6 | 1421 | |||
| GenBank mRNA | AK098661.1 | - | 4 | 423 | ||
| AK299691.1 | - | 4 | 698 | |||
| BC034033.1 | - | 5 | 960 | 4278547 | ||
| BX538061.1 | - | 4 | 513 | |||
| BX647386.1 | - | 5 | 765 | |||

CARNMT1
Genomic Map