siRNA Details
| Gene Symbol: PTPDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001253829.1 | NP_001240758.1 | 6 | 1372 | ||
| NM_001253830.1 | NP_001240759.1 | 7 | 1433 | |||
| NM_152422.4 | NP_689635.3 | 6 | 1366 | |||
| NM_177995.2 | NP_818931.1 | 7 | 1448 | |||
| XM_005251711.4 | XP_005251768.1 | 7 | 3390 | |||
| XM_011518240.2 | XP_011516542.1 | 7 | 1198 | |||
| XM_011518241.2 | XP_011516543.1 | 7 | 1152 | |||
| XM_017014280.1 | XP_016869769.1 | 8 | 1257 | |||
| XM_017014281.1 | XP_016869770.1 | 7 | 1176 | |||
| XM_017014282.1 | XP_016869771.1 | 4 | 1279 | |||
| GenBank mRNA | AK094631.1 | - | 1 | 118 | ||
| AK124912.1 | - | 6 | 1252 | |||
| AK289692.1 | - | 6 | 1323 | |||
| AK310403.1 | - | 6 | 1330 | |||
| AY171233.1 | - | 7 | 1453 | |||
| BC067120.1 | - | 7 | 1433 | 5539182 | ||

PTPDC1
Genomic Map