siRNA Details
| Gene Symbol: C9orf131 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040410.2 | NP_001035500.1 | 2 | 3134 | ||
| NM_001040411.2 | NP_001035501.1 | 2 | 3023 | |||
| NM_001040412.2 | NP_001035502.1 | 3 | 3098 | |||
| NM_001287391.1 | NP_001274320.1 | 2 | 3055 | |||
| NM_203299.3 | NP_976044.2 | 2 | 3239 | |||
| GenBank mRNA | AK097595.1 | - | 2 | 3205 | ||
| AK302123.1 | - | 3 | 1822 | |||
| AL133575.1 | - | 1 | 409 | |||
| BC045643.1 | - | 2 | 3218 | 5298387 | ||

C9orf131
Genomic Map