siRNA Details
| Gene Symbol: C9orf131 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040410.2 | NP_001035500.1 | 2 | 762 | ||
| NM_001040411.2 | NP_001035501.1 | 2 | 651 | |||
| NM_001040412.2 | NP_001035502.1 | 3 | 726 | |||
| NM_001287391.1 | NP_001274320.1 | 2 | 683 | |||
| NM_203299.3 | NP_976044.2 | 2 | 867 | |||
| GenBank mRNA | AK090398.1 | - | 1 | 875 | ||
| AK097595.1 | - | 2 | 833 | |||
| BC045643.1 | - | 2 | 846 | 5298387 | ||

C9orf131
Genomic Map