siRNA Details
| Gene Symbol: OR13C5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004482.1 | NP_001004482.1 | 1 | 734 | ||
| GenBank mRNA | BC136842.1 | - | 1 | 828 | 9020833 | |
| BC136843.1 | - | 1 | 828 | 9020834 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR13C5| Gene Symbol: OR13C5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004482.1 | NP_001004482.1 | 1 | 734 | ||
| GenBank mRNA | BC136842.1 | - | 1 | 828 | 9020833 | |
| BC136843.1 | - | 1 | 828 | 9020834 | ||