siRNA Details
| Gene Symbol: OR1L8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004454.1 | NP_001004454.1 | 1 | 915 | ||
| XM_017014284.1 | XP_016869773.1 | 3 | 1451 | |||
| XM_017014285.1 | XP_016869774.1 | 3 | 1451 | |||
| XM_017014286.1 | XP_016869775.1 | 2 | 1322 | |||
| GenBank mRNA | BC140910.1 | - | 1 | 997 | 9056722 | |

OR1L8
Genomic Map