siRNA Details
| Gene Symbol: SLITRK4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001184749.2 | NP_001171678.1 | 2 | 2605 | ||
| NM_001184750.2 | NP_001171679.1 | 2 | 2408 | |||
| NM_173078.4 | NP_775101.1 | 2 | 2558 | |||
| XM_005262365.4 | XP_005262422.1 | 2 | 2358 | |||
| XM_011531265.2 | XP_011529567.1 | 2 | 2403 | |||
| GenBank mRNA | AK057619.1 | - | 1 | 1275 | ||
| AK092016.1 | - | 2 | 2273 | |||
| AK315044.1 | - | 2 | 2408 | |||
| AL713693.1 | - | 1 | 1738 | |||
| BC040986.1 | - | 2 | 2567 | 4811695 | ||

SLITRK4
Genomic Map