siRNA Details
| Gene Symbol: FUNDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173794.3 | NP_776155.1 | 5 | 561 | ||
| GenBank mRNA | BC035015.2 | - | 5 | 445 | 4812334 | |
| BC042813.1 | - | 5 | 556 | 5274638 | ||
| BM837991.1 | - | 5 | 587 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FUNDC1| Gene Symbol: FUNDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173794.3 | NP_776155.1 | 5 | 561 | ||
| GenBank mRNA | BC035015.2 | - | 5 | 445 | 4812334 | |
| BC042813.1 | - | 5 | 556 | 5274638 | ||
| BM837991.1 | - | 5 | 587 | |||