siRNA Details
| Gene Symbol: ADGRG4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153834.3 | NP_722576.3 | 23 | 9033 | ||
| XM_011531269.2 | XP_011529571.1 | 22 | 9039 | |||
| XM_011531271.2 | XP_011529573.1 | 20 | 8771 | |||
| GenBank mRNA | AY140954.1 | - | 13 | 7986 | ||
| AY882585.1 | - | 23 | 9033 | |||
| CR936801.1 | - | 20 | 8277 | |||
More Information
Set Membership: |
|

ADGRG4
Genomic Map