siRNA Details
| Gene Symbol: DOCK11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144658.3 | NP_653259.3 | 45 | 5120 | ||
| XM_005262368.3 | XP_005262425.1 | 46 | 5404 | |||
| XM_005262369.4 | XP_005262426.1 | 46 | 5098 | |||
| XM_011531276.2 | XP_011529578.1 | 46 | 5039 | |||
| XM_011531278.2 | XP_011529580.1 | 33 | 3525 | |||
| GenBank mRNA | AK054649.1 | - | 6 | 802 | ||
| AK056684.1 | - | 15 | 1634 | |||
| AK124735.1 | - | 40 | 4543 | |||
| AK125641.1 | - | 24 | 2632 | |||
| AY692226.1 | - | 45 | 5120 | |||
| BC047713.1 | - | 11 | 1211 | |||

DOCK11
Genomic Map