siRNA Details
| Gene Symbol: ASB5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080874.3 | NP_543150.1 | 6 | 813 | ||
| XM_005262759.1 | XP_005262816.1 | 8 | 1242 | |||
| XM_011531616.2 | XP_011529918.1 | 6 | 921 | |||
| XM_011531617.2 | XP_011529919.1 | 6 | 1504 | |||
| GenBank mRNA | AK096441.1 | - | 6 | 715 | ||
| AK098693.1 | - | 6 | 718 | |||
| AY057053.1 | - | 6 | 699 | |||
| BC065710.1 | - | 6 | 720 | 4272258 | ||
| BX647857.1 | - | 6 | 814 | |||

ASB5
Genomic Map